Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Chicken IgG Fc Isotype Control

Click to view available options
:
1mg; Unlabeled
Description
IgG Fc Isotype Control antibody specifically detects IgG Fc in Chicken samples. It is validated for Western Blot, ELISA, Immunoprecipitation, SDS-Page.
Specifications
Specifications
For Use With (Application) | ELISA, Immunoprecipitation |
Formulation | Potassium Phosphate, 0.15M Sodium Chloride pH 7.2. |
Name | Chicken IgG, Fc Protein |
Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
Cross Reactivity | Chicken |
Species | Chicken |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction