Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Novus Biologicals™ Collagen XXIII alpha 1 Overexpression Lysate
 
    
    
    
        
            Shop All R&D Systems Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                0.1 mg
                            
                        
                    Description
The Collagen XXIII alpha 1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_775736. The protein contains a C-terminal DDK tag. | 
| For Use With (Application) | Western Blot | 
| Quantity | 0.1 mg | 
| Storage Buffer | RIPA buffer | 
| Purity | Protein | 
| Test Specificity | Homo sapiens COL23A1, mRNA. | 
| Content And Storage | -80°C. Avoid freeze-thaw cycles. | 
| Protein State | Native | 
For Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            