Learn More
Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC219651 and based on accession number NM_1879. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens mannan-binding lectin serine peptidase 1 (C4/C2 activating component of Ra-reactive factor) (MASP1), transcript variant 1, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Target | Complement Factor MASP1 |
| Synonym | 3MC1, C4/C2 activating component of Ra-reactive factor, Complement factor MASP-3, Complement-activating component of Ra-reactive factor, CRARF, CRARF1, DKFZp686I01199, FLJ26383, mannan binding lectin serine peptidase 1, mannan-binding lectin serine protease 1, Mannose-binding lectin-associated serine protease 1, Mannose-binding protein-associated serine protease, MAP1, MAP-1A, MAp44, MASP, MASP3, MASP-3, MGC126283, MGC126284, PRSS5, Ra-reactive factor serine protease p100, RaRF, Serine protease 5 |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
