Learn More
Description
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Digestive/Waste |
| Quantity | 4 Vials |
| Storage Buffer | 20mM Hepes (pH 7.5), 400mM NaCl, 20% glycerol, 0.1mM EDTA, 10mM NaF, 10μM Na2MoO4, 1mM NaVO3, 10mM PNPP, 10mM beta-glycerophosphate, 1mM DTT and protease inhibitors. |
| Purity | Protein |
| Test Specificity | NB800-PC26 contains hypoxia induced and uninduced COS-7 nuclear extract lysate |
| Cellular Fraction | Nuclear Hypoxic Induced |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
