Learn More
Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC24171 and based on accession number NM_1356. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked (DDX3X), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Target | DDX3X |
| Synonym | ATP-dependent RNA helicase DDX3X, CAP-Rf, DBX, DDX14, DDX3, DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, DEAD/H box-3, EC 3.6.4.13, Helicase-like protein 2, HLP2, MRX102, MRXSSB |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
