Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DFF45/ICAD Antibody Blocking Peptide
SDP

Catalog No. NBP177010E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177010E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177010E Supplier Novus Biologicals™ Supplier No. NBP177010PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 20 amino amino terminus of mouse ICAD. Source: Synthetic (Accession #: O54786) The DFF45/ICAD Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 1676
Species Mouse
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-77010. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias DFF1DNA fragmentation factor 45 kDa subunit, DFF45DNA fragmentation factor subunit alpha, DFF-45 DNA fragmentation factor, 45 kD, alpha polypeptide, DNA fragmentation factor, 45 kDa, alpha polypeptide, ICAD, Inhibitor of CAD
Gene Symbol DFFA
Product Type DFF45/ICAD
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination 20 amino amino terminus of mouse ICAD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.