Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DIRAS1 Protein
SDP

Catalog No. NBP14859705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14859705 0.05 mg
NBP14859702 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14859705 Supplier Novus Biologicals™ Supplier No. NBP1485970.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-195 of Human DIRAS1 The Recombinant Human DIRAS1 Protein is derived from E. coli. The Recombinant Human DIRAS1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1mM DTT, 0.1M NaCl, 1mM EDTA.
Gene ID (Entrez) 148252
Molecular Weight (g/mol) 24.1kDa
Purification Method Protein
Quantity 0.05 mg
Source Human
Immunogen DIRAS1, 1-195 aa. Sequence: MGSSHHHHHHSSGLVPRGSHMPEQSNDYRVVVFGAGGVGKSSLVLRFVKGTFRDTYIPTIEDTYRQVISCDKSVCTLQITDTTGSHQFPAMQRLSISKGHAFILVFSVTSKQSLEELGPIYKLIVQIKGSVEDIPVMLVGNKCDETQREVDTREAQAVAQEWKCAFMETSAKMNYNVKELFQELLTLETRRNMSLNIDGKRSGKQKRTDRVKGKC MGSSHHHHHHSSGLVPRGSHMPEQSNDYRVVVFGAGGVGKSSLVLRFVKGTFRDTYIPTIEDTYRQVISCDKSVCTLQITDTTGSHQFPAMQRLSISKGHAFILVFSVTSKQSLEELGPIYKLIVQIKGSVEDIPVMLVGNKCDETQREVDTREAQAVAQEWKCAFMETSAKMNYNVKELFQELLTLETRRNMSLNIDGKRSGKQKRTDRVKGKC
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name DIRAS1
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.