Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 1644 |
| Species | Human |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized from sterile distilled water. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-56918. This Synthetic Peptide is Designed for Use in an Antibody Competition Assay with Its Corresponding Antibody. Use of This Product in Any Other Assay Has Not Yet Been Tested. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28 |
| Gene Symbol | DDC |
| Molecular Weight (g/mol) | 53kDa |
| Product Type | Dopa Decarboxylase/DDC |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
