Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ eIF4E Protein
SDP

Catalog No. NBP14531405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
0.05 mg
0.25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14531405 0.05 mg
NBP14531402 0.25 mg
NB398565 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14531405 Supplier Novus Biologicals™ Supplier No. NBP1453140.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-217 of Human eIF4E The Recombinant Human eIF4E Protein is derived from E. coli. The Recombinant Human eIF4E Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001959
Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol
Gene ID (Entrez) 1977
Molecular Weight (g/mol) 27.2kDa
Purification Method Protein
Quantity 0.05 mg
Source Recombinant human EIF4E protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen EIF4E, 1-217aa, Human, His tag, E. coli (NP_001959).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name eIF4E
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.