Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EMI1 Antibody Blocking Peptide
SDP

Catalog No. NBP196758 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196758 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196758 Supplier Novus Biologicals™ Supplier No. NBP196758
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from EMI1 Source: Synthetic The EMI1 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 26271
Species Human
Purity >90%
Accession Number Q9UKT4
Content And Storage Store at -20 C short term. Aliquot and store (at -80 C long term. Avoid freeze-thaw cycles.)
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
For Use With (Application) This product should be used at 1.0μg per 1.0μL of antiserum in per assay.
Gene Alias EMI1Early mitotic inhibitor 1, F-box only protein 5, F-box protein 5, FBX5F-box protein Fbx5, Fbxo31
Gene Symbol FBXO5
Product Type EMI1 Peptide
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination Sequence: Ser - Gly - Tyr - Glu - Val - His - His - Gln - Lys - Leu - Val - Phe - Phe -OH (aa 8-20)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.