Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FDX1 Protein
SDP

Catalog No. NBP1508295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1508295 0.5 mg
NBP15082901 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1508295 Supplier Novus Biologicals™ Supplier No. NBP1508290.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal T7-tag and corresponding to the amino acids 61-184 of Human FDX1 The Recombinant Human FDX1 Protein is derived from E. coli. The Recombinant Human FDX1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004100
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% Glycerol
Gene ID (Entrez) 2230
Molecular Weight (g/mol) 15.0kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Recombinant human Ferredoxin 1 protein, fused to T7-tag at N-terminus, expressed in E. coli
Immunogen Ferredoxin 1, 61-184 aa, Human, T7-tagged, Recombinant, E. coli (NP_004100).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name FDX1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.