Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Fibrinogen beta chain Overexpression Lysate
SDP

Catalog No. NBL110684 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBL110684 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBL110684 Supplier Novus Biologicals™ Supplier No. NBL110684
Add to Cart
Add to Cart
The Fibrinopeptide B Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC21471 and based on accession number NM_5141. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens fibrinogen beta chain (FGB), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Target Fibrinogen beta chain
Synonym Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78 p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78 p, MGC104327, MGC120405
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.