Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ gamma Tubulin Protein
SDP

Catalog No. NBP17253410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17253410 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17253410 Supplier Novus Biologicals™ Supplier No. NBP172534100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-451 of Human gamma Tubulin The Recombinant Human gamma Tubulin Protein is derived from E. coli. The Recombinant Human gamma Tubulin Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1.0mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) 1M Urea, 5% Glycerol.
Molecular Weight (g/mol) 53.3kDa
Purification Method SDS-PAGE
Quantity 100 μg
Source Recombinant human TUBG1 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen TUBG1, 1-451 aa, Human, His tag N terminus E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name gamma Tubulin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.