Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HeLa CoCl2 treated/Untreated Cell Lysate
SDP

Catalog No. NBP236450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236450 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236450 Supplier Novus Biologicals™ Supplier No. NBP236450
Add to Cart
Add to Cart
The HeLa Hypoxic (CoCl2)/ Normoxic Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.

Specifications

Tissue Cervix
Quantity 2 Vials
Storage Buffer The protein lysate is prepared in 1X Laemmli sample buffer with BME. Boiling before loading is not necessary.
Cellular Fraction Hypoxic (CoCl2) / Normoxic
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.