Learn More
Description
Specifications
Specifications
| Quantity | 500 μg |
| Storage Buffer | 1X SDS-PAGE Sample Buffer [0.05mM Tris HCl, 2% SDS, 10% Glycerol, 0.1% bromophenol blue, pH 6.8] |
| Test Specificity | Hep2 whole cell extract lysate |
| Cellular Fraction | Whole |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| For Use With (Application) | Western Blot |
| Tissue | Reproductive |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
