Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Histone H3 [Dimethyl Lys36] Antibody Blocking Peptide
SDP

Catalog No. NB211252PEP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.04 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211252PEP 0.04 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211252PEP Supplier Novus Biologicals™ Supplier No. NB211252PEP
Add to Cart
Add to Cart
A peptide to HIST2H3C.

Specifications

Gene ID (Entrez) 126961
Species Human
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Lyophilized. Reconstitute in 40 ul of diH20.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB21-1252. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias H3, H3 histone, family 3 A, H3.3 A, H3.3 B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Gene Symbol HIST2H3C
Product Type Histone H3
Quantity 0.04 mg
Regulatory Status RUO - research use only
Research Category Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.