Learn More
Description
Specifications
Specifications
| For Use With (Application) | ELISA, Immunoassay |
| Formulation | Sterile liquid; 20mM Tris-HCl, pH 8.0, 0.3M NaCl, 1mM DTT. 0.22μm filtered. |
| Quantity | 0.5 mg |
| Source | Mouse |
| Immunogen | HMGB1/HMG-1 |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | The recombinant mouse HMGB-1 has been tested by bioassay, yielding ED50 of 3μg/mL. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
