Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HSP70/HSPA1A Protein
SDP

Catalog No. NBC1183675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC1183675 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC1183675 Supplier Novus Biologicals™ Supplier No. NBC1183670.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-641 of Human HSP70/HSPA1A The Recombinant Human HSP70/HSPA1A Protein is derived from E. coli. The Recombinant Human HSP70/HSPA1A Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_005336
Concentration 1.0mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris pH 7.5, 2mM DTT
Gene ID (Entrez) 3303
Purification Method Protein
Quantity 0.5 mg
Source Recombinant human Hsp70, fused to His-tag at N-terminus, expressed in E. coli
Immunogen HSP70 (amino acids 1-641) Human, Recombinant, His- tagged, E. coli (NP_005336).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name HSP70/HSPA1A
Conjugate Unconjugated
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.