Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Apolipoprotein L4 His Protein
SDP

Catalog No. NBP222692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222692 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222692 Supplier Novus Biologicals™ Supplier No. NBP222692
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-348 of Human Apolipoprotein L4 The Recombinant Human Apolipoprotein L4 Protein is derived from E. coli. The Recombinant Human Apolipoprotein L4 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_085146
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 80832
Molecular Weight (g/mol) 41.1kDa
Name Apolipoprotein L4 Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen APOL4, 1-348aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMGSWVQL ITSVGVQQNH PGWTVAGQFQ EKKRFTEEVI EYFQKKVSPV HLKILLTSDE AWKRFVRVAE LPREEADALY EALKNLTPYV AIEDKDMQQK EQQFREWFLK EFPQIRWKIQ ESIERLRVIA NEIEKVHRGC VIANVVSGST GILSVIGVML APFTAGLSLS ITAAGVGLGI ASATAGIASS IVENTYTRSA ELTASRLTAT STDQLEALRD ILRDITPNVL SFALDFDEAT KMIANDVHTL RRSKATVGRP LIAWRYVPIN VVETLRTRGA PTRIVRKVAR NLGKATSGVL VVLDVVNLVQ DSLDLHKGAK SESAESLRQW AQELEENLNE LTHIHQSLKA G
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.