Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ASNA1 His Protein
SDP

Catalog No. NBP222694 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222694 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222694 Supplier Novus Biologicals™ Supplier No. NBP222694
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-348 of Human ASNA1 The Recombinant Human ASNA1 Protein is derived from E. coli. The Recombinant Human ASNA1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004308
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 439
Molecular Weight (g/mol) 41.2kDa
Name ASNA1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen ASNA1, 1-348aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAAGVAG WGVEAEEFED APDVEPLEPT LSNIIEQRSL KWIFVGGKGG VGKTTCSCSL AVQLSKGRES VLIISTDPAH NISDAFDQKF SKVPTKVKGY DNLFAMEIDP SLGVAELPDE FFEEDNMLSM GKKMMQEAMS AFPGIDEAMS YAEVMRLVKG MNFSVVVFDT APTGHTLRLL NFPTIVERGL GRLMQIKNQI SPFISQMCNM LGLGDMNADQ LASKLEETLP VIRSVSEQFK DPEQTTFICV CIAEFLSLYE TERLIQELAK CKIDTHNIIV NQLVFPDPEK PCKMCEARHK IQAKYLDQME DLYEDFHIVK LPLLPHEVRG ADKVNTFSAL LLEPYKPPSA Q
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol GET3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.