Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ATP5H His Protein
SDP

Catalog No. NBP222700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222700 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222700 Supplier Novus Biologicals™ Supplier No. NBP222700
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-161 of Human ATP5H The Recombinant Human ATP5H Protein is derived from E. coli. The Recombinant Human ATP5H Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_006347
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 10476
Molecular Weight (g/mol) 20.9kDa
Name ATP5H Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen ATP5H, 1-161aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAGRKLA LKTIDWVAFA EIIPQNQKAI ASSLKSWNET LTSRLAALPE NPPAIDWAYY KANVAKAGLV DDFEKKFNAL KVPVPEDKYT AQVDAEEKED VKSCAEWVSL SKARIVEYEK EMEKMKNLIP FDQMTIEDLN EAFPETKLDK KKYPYWPHQP IENL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol ATP5PD
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.