Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Bcl 7A His Protein
SDP

Catalog No. NBP222707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222707 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222707 Supplier Novus Biologicals™ Supplier No. NBP222707
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-210 of Human Bcl 7A The Recombinant Human Bcl 7A Protein is derived from E. coli. The Recombinant Human Bcl 7A Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001019979
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT, 2mM EDTA
Gene ID (Entrez) 605
Molecular Weight (g/mol) 25.2kDa
Name Bcl 7A Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen BCL7A, 1-210aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSGRSVR AETRSRAKDD IKRVMAAIEK VRKWEKKWVT VGDTSLRIYK WVPVTEPKVD DKNKNKKKGK DEKCGSEVTT PENSSSPGMM DMHDDNSNQS SIADASPIKQ ENSSNSSPAP EPNSAVPSDG TEAKVDEAQA DGKEHPGAED ASDEQNSQSS MEHSMNSSEK VDRQPSGDSG LAAETSAISQ DLEGVPPSKK MKLEASQQNS EEM
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.