Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human BOLA1 His Protein
SDP

Catalog No. NBP222714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222714 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222714 Supplier Novus Biologicals™ Supplier No. NBP222714
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 21-137 of Human BOLA1 The Recombinant Human BOLA1 Protein is derived from E. coli. The Recombinant Human BOLA1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_057158
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 51027
Molecular Weight (g/mol) 14kDa
Name BOLA1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen BOLA1, 21-137aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSQGSAGSG AIGPVEAAIR TKLEEALSPE VLELRNESGG HAVPPGSETH FRVAVVSSRF EGLSPLQRHR LVHAALAEEL GGPVHALAIQ ARTPAQWREN SQLDTSPPCL GGNKKTLGTP
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.