Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CHMP1a His Protein
SDP

Catalog No. NBP222770 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222770 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222770 Supplier Novus Biologicals™ Supplier No. NBP222770
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-196 of Human CHMP1a The Recombinant Human CHMP1a Protein is derived from E. coli. The Recombinant Human CHMP1a Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002759
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 5119
Molecular Weight (g/mol) 24.1kDa
Name CHMP1a Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen CHMP1A, 1-196aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMDDTLFQ LKFTAKQLEK LAKKAEKDSK AEQAKVKKAL LQKNVECARV YAENAIRKKN EGVNWLRMAS RVDAVASKVQ TAVTMKGVTK NMAQVTKALD KALSTMDLQK VSSVMDRFEQ QVQNLDVHTS VMEDSMSSAT TLTTPQEQVD SLIMQIAEEN GLEVLDQLSQ LPEGASAVGE SSVRSQEDQL SRRLAALRN
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.