Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CIB1 His Protein
SDP

Catalog No. NBC118394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118394 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118394 Supplier Novus Biologicals™ Supplier No. NBC118394
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-191 of Human CIB1 The Recombinant Human CIB1 Protein is derived from E. coli. The Recombinant Human CIB1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_006375
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 5 mM DTT
Gene ID (Entrez) 10519
Molecular Weight (g/mol) M.W. (theoretical): 23 kDa
Name CIB1 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHHSSGLVPRGSHMGGSGSRLSKELLAEYQDLTFLTKQEILLAHRRFCELLPQEQRTVESSLRAQVPFEQILSLPELKANPFKERICRVFSTSPAKDSLSFEDFLDLLSVFSDTATPDIKSHYAFRIFDFDDDGTLNREDLSRLVNCLTGEGEDTRLSASEMKQLIDNILEESDIDRDGTINLSEFQHVISRSPDFASSFKIVL
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol CIB1
Cross Reactivity Human
Research Category Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE
Protein CIB1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.