Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CKS2 T7 Protein
SDP

Catalog No. NBC118431 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118431 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118431 Supplier Novus Biologicals™ Supplier No. NBC118431
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal T7-tag and corresponding to the amino acids 1-79 of Human CKS2 The Recombinant Human CKS2 Protein is derived from E. coli. The Recombinant Human CKS2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001818
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris buffer (pH 7.5), 20% Glycerol
Gene ID (Entrez) 1164
Molecular Weight (g/mol) 11.3kDa
Name CKS2 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MASMTGGQQMGRGSHMAHKQIYYSDKYFDEHYEYRHVMLPRELSKQVPKTHLMSEEEWRRLGVQQSLGWVHYMIHEPEPHILLFRRPLPKDQQK
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.