Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CUEDC2 His Protein
SDP

Catalog No. NBP222968 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222968 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222968 Supplier Novus Biologicals™ Supplier No. NBP222968
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-287 of Human CUEDC2 The Recombinant Human CUEDC2 Protein is derived from E. coli. The Recombinant Human CUEDC2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_076945
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 79004
Molecular Weight (g/mol) theoretical_molecular_weight: L606
Name CUEDC2 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen CUEDC2, 1-287aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMELERIV SAALLAFVQT HLPEADLSGL DEVIFSYVLG VLEDLGPSGP SEENFDMEAF TEMMEAYVPG FAHIPRGTIG DMMQKLSGQL SDARNKENLQ PQSSGVQGQV PISPEPLQRP EMLKEETRSS AAAAADTQDE ATGAEEELLP GVDVLLEVFP TCSVEQAQWV LAKARGDLEE AVQMLVEGKE EGPAAWEGPN QDLPRRLRGP QKDELKSFIL QKYMMVDSAE DQKIHRPMAP KEAPKKLIRY IDNQVVSTKG ERFKDVRNPE AEEMKATYIN LKPARKYRFH
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.