Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Dihydrofolate Reductase/DHFR His Protein
SDP

Catalog No. NBP130207 Shop All R&D Systems Products
Change view
Click to view available options
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP130207 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130207 Supplier Novus Biologicals™ Supplier No. NBP130207

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-187 of Human Dihydrofolate Reductase/DHFR The Recombinant Human Dihydrofolate Reductase/DHFR Protein is derived from E. coli. The Recombinant Human Dihydrofolate Reductase/DHFR Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number P00374
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 0.1 M Nacl 2mM DTT, and 30% glycerol
Gene ID (Entrez) 1719
Molecular Weight (g/mol) 23.6kDa
Name Dihydrofolate Reductase/DHFR Protein
Purification Method Protein
Quantity 0.1mg
Source E. Coli
Immunogen DHFR, 1-187 aa, Human, His-tagged, Recombinant, E.coli (NP_000782.1).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.