Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Dysbindin His Protein
SDP

Catalog No. NBC121266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC121266 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC121266 Supplier Novus Biologicals™ Supplier No. NBC121266
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-270 of Human Dysbindin The Recombinant Human Dysbindin Protein is derived from E. coli. The Recombinant Human Dysbindin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_898862
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 0.5mM DTT, 20% glycerol
Gene ID (Entrez) 84062
Molecular Weight (g/mol) 34.6kDa
Name Dysbindin Protein
Purification Method Affinity purified
Quantity 0.1 mg
Immunogen MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMLS AHWEKKKTSL VELQEQLQQL PALIADLESM TANLTHLEAS FEEVENNLLH LEDLCGQCEL ERCKHMQSQQ LENYKKNKRK ELETFKAELD AEHAQKVLEM EHTQQMKLKE RQKFFEEAFQ QDMEQYLSTG YLQIAERREP IGSMSSMEVN VDMLEQMDLM DISDQEALDV FLNSGGEENT VLSPALGPES STCQNEITLQ VPNPSELRAK PPSSSSTCTD SATRDISEGG ESPVVQSDEE EVQVDTALAT SHTDREATPD GGEDSDS
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Research Category Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.