Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human EIF3F His Protein
SDP

Catalog No. NBP223030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223030 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223030 Supplier Novus Biologicals™ Supplier No. NBP223030
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-357 of Human EIF3F The Recombinant Human EIF3F Protein is derived from E. coli. The Recombinant Human EIF3F Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003745
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 8665
Molecular Weight (g/mol) 400kDa
Name EIF3F Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen EIF3F, 1-357 aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMATPAVP VSAPPATPTP VPAAAPASVP APTPAPAAAP VPAAAPASSS DPAAAAAATA APGQTPASAQ APAQTPAPAL PGPALPGPFP GGRVVRLHPV ILASIVDSYE RRNEGAARVI GTLLGTVDKH SVEVTNCFSV PHNESEDEVA VDMEFAKNMY ELHKKVSPNE LILGWYATGH DITEHSVLIH EYYSREAPNP IHLTVDTSLQ NGRMSIKAYV STLMGVPGRT MGVMFTPLTV KYAYYDTERI GVDLIMKTCF SPNRVIGLSS DLQQVGGASA RIQDALSTVL QYAEDVLSGK VSADNTVGRF LMSLVNQVPK IVPDDFETML NSNINDLLMV TYLANLTQSQ IALNEKLVNL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.