Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Eukaryotic translation initiation factor 1 His Protein
SDP

Catalog No. NBP137080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP137080 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP137080 Supplier Novus Biologicals™ Supplier No. NBP137080
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-113 of Human Eukaryotic translation initiation factor 1 The Recombinant Human Eukaryotic translation initiation factor 1 Protein is derived from E. coli. The Recombinant Human Eukaryotic translation initiation factor 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_005792
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 0.1 M NaCl, and 10% glycerol
Gene ID (Entrez) 10209
Molecular Weight (g/mol) 16.9kDa
Name Eukaryotic translation initiation factor 1 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWAGSMSA IQNLHSFDPF ADASKGDDLL PAGTEDYIHI RIQQRNGRKT LTTVQGIADD YDKKKLVKAF KKKFACNGTV IEHPEYGEVI QLQGDQRKNI CQFLVEIGLA KDDQLKVHGF
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Research Category DNA replication Transcription Translation and Splicing
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.