Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Exosome component 4 His Protein
SDP

Catalog No. NBP223032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223032 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223032 Supplier Novus Biologicals™ Supplier No. NBP223032
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-245 of Human Exosome component 4 The Recombinant Human Exosome component 4 Protein is derived from E. coli. The Recombinant Human Exosome component 4 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_061910
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 54512
Molecular Weight (g/mol) 28.8kDa
Name Exosome component 4 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen exosome component 4 Protein, 1-245aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAGLELL SDQGYRVDGR RAGELRKIQA RMGVFAQADG SAYIEQGNTK ALAVVYGPHE IRGSRARALP DRALVNCQYS SATFSTGERK RRPHGDRKSC EMGLQLRQTF EAAILTQLHP RSQIDIYVQV LQADGGTYAA CVNAATLAVL DAGIPMRDFV CACSAGFVDG TALADLSHVE EAAGGPQLAL ALLPASGQIA LLEMDARLHE DHLERVLEAA AQAARDVHTL LDRVVRQHVR EASILLGD
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.