Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Fructosamine-3-kinase-related His Protein
SDP

Catalog No. NBP223060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223060 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223060 Supplier Novus Biologicals™ Supplier No. NBP223060
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-309 of Human Fructosamine-3-kinase-related The Recombinant Human Fructosamine-3-kinase-related Protein is derived from E. coli. The Recombinant Human Fructosamine-3-kinase-related Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_078895
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 79672
Molecular Weight (g/mol) 36.8kDa
Name Fructosamine-3-kinase-related Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen FN3KRP, 1-309aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMEELLRR ELGCSSVRAT GHSGGGCISQ GRSYDTDQGR VFVKVNPKAE ARRMFEGEMA SLTAILKTNT VKVPKPIKVL DAPGGGSVLV MEHMDMRHLS SHAAKLGAQL ADLHLDNKKL GEMRLKEAGT VGRGGGQEER PFVARFGFDV VTCCGYLPQV NDWQEDWVVF YARQRIQPQM DMVEKESGDR EALQLWSALQ LKIPDLFRDL EIIPALLHGD LWGGNVAEDS SGPVIFDPAS FYGHSEYELA IAGMFGGFSS SFYSAYHGKI PKAPGFEKRL QLYQLFHYLN HWNHFGSGYR GSSLNIMRNL VK
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.