Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GAPDH Protein
SDP

Catalog No. NBC118528
Click to view available options
Quantity:
0.1 mg

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-335 of Human GAPDH The Recombinant Human GAPDH Protein is derived from E. coli. The Recombinant Human GAPDH Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P04406
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 1 mM EDTA, 1 mM DTT, 20% glycerol
Gene ID (Entrez) 2597
Molecular Weight (g/mol) M.W. (theoretical): 36 kDa
Name GAPDH Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen Human GAPDH, 1-335 aa (NP_002037).
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbol GAPDH
Cross Reactivity Human
Research Category Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Purity or Quality Grade >95%, by SDS-PAGE
Protein GAPDH
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.