Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Glutathione S-Transferase mu 1/GSTM1 Recombinant Protein
SDP

Catalog No. NBC119065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC119065 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC119065 Supplier Novus Biologicals™ Supplier No. NBC119065

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-218 of Mouse Glutathione S-Transferase mu 1/GSTM1 The Recombinant Mouse Glutathione S-Transferase mu 1/GSTM1 Protein is derived from E. coli. The Recombinant Mouse Glutathione S-Transferase mu 1/GSTM1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4), 5mM glutathione
Gene ID (Entrez) 2944
Molecular Weight (g/mol) 26kDa
Name Glutathione S-Transferase mu 1/GSTM1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen MPMILGYWNV RGLTHPIRML LEYTDSSYDE KRYTMGDAPD FDRSQWLNEK FKLGLDFPNL PYLIDGSHKI TQSNAILRYL ARKHHLDGET EEERIRADIV ENQVMDTRMQ LIMLCYNPDF EKQKPEFLKT IPEKMKLYSE FLGKRPWFAG DKVTYVDFLA YDILDQYRMF EPKCLDAFPN LRDFLARFEG LKKISAYMKS SRYIATPIFS KMAHWSNK
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.