Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GSTZ1 His Protein
SDP

Catalog No. NBP140410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP140410 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140410 Supplier Novus Biologicals™ Supplier No. NBP140410
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-216 of Human GSTZ1 The Recombinant Human GSTZ1 Protein is derived from E. coli. The Recombinant Human GSTZ1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number AAC33591
For Use With (Application) ELISA, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 2954
Molecular Weight (g/mol) 26.2kDa
Name GSTZ1 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MQAGKPILYS YFRSSCSWRV RIALALKGID YETVPINLIK DGGQQFSKDF QALNPMKQVP TLKIDGITIH QSLAIIEYLE ETRPTPRLLP QDPKKRASVR MISDLIAGGI QPLQNLSVLK QVGEEMQLTW AQNAITCGFN ALEQILQSTA GIYCVGDEVT MADLCLVPQV ANAERFKVDL TPYPTISSIN KRLLVLEAFQ VSHPCRQPDT PTELRA
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Research Category metabolism
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.