Learn More
Description
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Western Blot |
| Formulation | 50mM Tris pH 8.0, 200mM NaCl, 8 M Urea, 10mM Imidazole |
| Gene ID (Entrez) | 3014 |
| Name | Histone H2AX |
| Quantity | 10 μg |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Gene Alias | H2A.X, H2A/X, H2AFX |
| Cross Reactivity | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
