Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Hexokinase 2 His Protein
SDP

Catalog No. NBC118389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118389 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118389 Supplier Novus Biologicals™ Supplier No. NBC118389

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Specific activity is > 12,000 pmol/min/ug. One unit will convert 1 pmole of D-Glucose to D-Glucose-6-phosphate per minute at pH8.0 at 37C.

Specifications

Accession Number P52789
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) 10% glycerol
Gene ID (Entrez) 3099
Molecular Weight (g/mol) 104.1kDa
Name Hexokinase 2 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Hexokinase 2 Human, His-tagged, Recombinant, E.coli (NP_000180).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.