Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HINT2 His Protein
SDP

Catalog No. NBP223124 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223124 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223124 Supplier Novus Biologicals™ Supplier No. NBP223124
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 18-163 of Human HINT2 The Recombinant Human HINT2 Protein is derived from E. coli. The Recombinant Human HINT2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_115982
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 84681
Molecular Weight (g/mol) 17.9kDa
Name HINT2 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen HINT2, 18-163aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSVAATGVR GGQVRGAAGV TDGNEVAKAQ QATPGGAAPT IFSRILDKSL PADILYEDQQ CLVFRDVAPQ APVHFLVIPK KPIPRISQAE EEDQQLLGHL LLVAKQTAKA EGLGDGYRLV INDGKLGAQS VYHLHIHVLG GRQLQWPPG
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.