Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human hnRNP AB His Protein
SDP

Catalog No. NBP223119 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223119 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223119 Supplier Novus Biologicals™ Supplier No. NBP223119
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-285 of Human hnRNP AB The Recombinant Human hnRNP AB Protein is derived from E. coli. The Recombinant Human hnRNP AB Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004490
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 30% glycerol, 1mM DTT
Gene ID (Entrez) 3182
Molecular Weight (g/mol) 33kDa
Name hnRNP AB Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen HNRNPAB, 1-285aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSEAGEE QPMETTGATE NGHEAVPEGE SPAGAGTGAA AGAGGATAAP PSGNQNGAEG DQINASKNEE DAGKMFVGGL SWDTSKKDLK DYFTKFGEVV DCTIKMDPNT GRSRGFGFIL FKDAASVEKV LDQKEHRLDG RVIDPKKAMA MKKDPVKKIF VGGLNPEATE EKIREYFGEF GEIEAIELPM DPKLNKRRGF VFITFKEEEP VKKVLEKKFH TVSGSKCEIK VAQPKEVYQQ QQYGSGGRGN RNRGNRGSGG GGGGGGQGST NYGKSQRRGG HQNNYKPY
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.