Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HspA6 His Protein
SDP

Catalog No. NBP130291 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130291 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130291 Supplier Novus Biologicals™ Supplier No. NBP130291
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-643 of Human HspA6 The Recombinant Human HspA6 Protein is derived from E. coli. The Recombinant Human HspA6 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002146
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 10% glycerol
Gene ID (Entrez) 3310
Molecular Weight (g/mol) 73.2kDa
Name HspA6 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MQAPRELAVG IDLGTTYSCV GVFQQGRVEI LANDQGNRTT PSYVAFTDTE RLVGDAAKSQ AALNPHNTVF DAKRLIGRKF ADTTVQSDMK HWPFRVVSEG GKPKVRVCYR GEDKTFYPEE ISSMVLSKMK ETAEAYLGQP VKHAVITVPA YFNDSQRQAT KDAGAIAGLN VLRIINEPTA AAIAYGLDRR GAGERNVLIF DLGGGTFDVS VLSIDAGVFE VKATAGDTHL GGEDFDNRLV NHFMEEFRRK HGKDLSGNKR ALRRLRTACE RAKRTLSSST QATLEIDSLF EGVDFYTSIT RARFEELCSD LFRSTLEPVE KALRDAKLDK AQIHDVVLVG GSTRIPKVQK LLQDFFNGKE LNKSINPDEA VAYGAAVQAA VLMGDKCEKV QDLLLLDVAP LSLGLETAGG VMTTLIQRNA TIPTKQTQTF TTYSDNQPGV FIQVYEGERA MTKDNNLLGR FELSGIPPAP RGVPQIEVTF DIDANGILSV TATDRSTGKA NKITITNDKG RLSKEEVERM VHEAEQYKAE DEAQRDRVAA KNSLEAHVFH VKGSLQEESL RDKIPEEDRR KMQDKCREVL AWLEHNQLAE KEEYEHQKRE LEQICRPIFS RLYGGPGVPG GSSCGTQARQ GDPSTGPIIE EVD
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.