Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HSPB8/HSP22 His Protein
SDP

Catalog No. NBP141159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141159 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141159 Supplier Novus Biologicals™ Supplier No. NBP141159
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-196 of Human HSPB8/HSP22 The Recombinant Human HSPB8/HSP22 Protein is derived from E. coli. The Recombinant Human HSPB8/HSP22 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_055180
Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 100 mM NaCl, 20% glycerol
Gene ID (Entrez) 26353
Molecular Weight (g/mol) M.W. (theoretical): 23.7 kDa
Name HSPB8/HSP22 Protein
Purification Method Protein
Quantity 0.05 mg
Immunogen MGSSHHHHHH SSGLVPRGSH MADGQMPFSC HYPSRLRRDP FRDSPLSSRL LDDGFGMDPF PDDLTASWPD WALPRLSSAW PGTLRSGMVP RGPTATARFG VPAEGRTPPP FPGEPWKVCV NVHSFKPEEL MVKTKDGYVE VSGKHEEKQQ EGGIVSKNFT KKIQLPAEVD PVTVFASLSP EGLLIIEAPQ VPPYSTFGES SFNNELPQDS QEVTCT
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Symbol HSPB8
Cross Reactivity Human
Research Category Protein Kinase
Purity or Quality Grade >90%, by SDS-PAGE
Protein HSPB8/HSP22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.