Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HspBP1 His Protein
SDP

Catalog No. NBP144481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144481 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144481 Supplier Novus Biologicals™ Supplier No. NBP144481
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-362 of Human HspBP1 The Recombinant Human HspBP1 Protein is derived from E. coli. The Recombinant Human HspBP1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number AAH01236
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 30% glycerol, 2mM EDTA, 0.1 M NaCl
Gene ID (Entrez) 23640
Molecular Weight (g/mol) 41.6kDa
Name HspBP1 Protein
Purification Method Protein
Quantity 50 μg
Immunogen MGSSHHHHHH SSGLVPRGSH MSDEGSRGSR LPLALPPASQ GCSSGGGGGG GGGSSAGGSG NSRPPRNLQG LLQMAITAGS EEPDPPPEPM SEERRQWLQE AMSAAFRGQR EEVEQMKSCL RVLSQPMPPT AGEAEQAADQ QEREGALELL ADLCENMDNA ADFCQLSGMH LLVGRYLEAG AAGLRWRAAQ LIGTCSQNVA AIQEQVLGLG ALRKLLRLLD RDACDTVRVK ALFAISCLVR EQEAGLLQFL RLDGFSVLMR AMQQQVQKLK VKSAFLLQNL LVGHPEHKGT LCSMGMVQQL VALVRTEHSP FHEHVLGALC SLVTDFPQGV RECREPELGL EELLRHRCQL LQQHEEYQEE LEFCEKLLQT CFSSPADDSM DR
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.