Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ICT His Protein
SDP

Catalog No. NBP223129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223129 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223129 Supplier Novus Biologicals™ Supplier No. NBP223129
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 30-206 of Human ICT The Recombinant Human ICT Protein is derived from E. coli. The Recombinant Human ICT Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001536
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 30% glycerol, 1mM DTT
Gene ID (Entrez) 3396
Molecular Weight (g/mol) 22.8kDa
Name ICT Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen ICT1, 30-206aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSLHKQKDG TEFKSIYSLD KLYPESQGSD TAWRVPNGAK QADSDIPLDR LTISYCRSSG PGGQNVNKVN SKAEVRFHLA TAEWIAEPVR QKIAITHKNK INRLGELILT SESSRYQFRN LADCLQKIRD MITEASQTPK EPTKEDVKLH RIRIENMNRE RLRQKRIHSA VKTSRRVDMD
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol MRPL58
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Cancer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.