Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-1 beta/IL-1F2 Protein
SDP

Catalog No. NBC118478 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118478 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118478 Supplier Novus Biologicals™ Supplier No. NBC118478

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Human IL-1 beta/IL-1F2 stimulates cell proliferation of the D10.G4.1 mouse helper T cells. The ED50 range is less or equal 12 pg/ml.

Specifications

Accession Number P01584
Concentration 1 mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4)
Gene ID (Entrez) 3553
Molecular Weight (g/mol) M.W. (theoretical): 17 kDa
Name IL-1 beta/IL-1F2 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen IL-1beta, 117-269aa, Human, Recombinant, E.coli (NP_000567).
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of protein (determined by LAL method)
Gene Symbol IL1B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Adaptive Immunity, Apoptosis, Cancer, Cytokine Research, Diabetes Research, Immunology, Innate Immunity
Purity or Quality Grade >95%, by SDS-PAGE
Protein IL-1 beta/IL-1F2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.