Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human IL-12 Protein

Click to view available options
:
0.01mg; Unlabeled
Description
Recombinant bioactive protein for Human IL-12 The Recombinant Human IL-12 Protein is derived from CHO. The Recombinant Human IL-12 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Accession Number | P29459 |
Formulation | Sterile filtered and lyophilized in 0.5X PBS, pH 7.2. |
Gene ID (Entrez) | 3592 |
Name | IL-12 Protein |
Purification Method | SDS-PAGE |
Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Purity or Quality Grade | >98%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction