Learn More
Description
Specifications
Specifications
| Accession Number | NP_000577 |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Formulation | 20mM MES buffer (pH5.0), 10% glycerol |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | 15.4kDa |
| Name | IL-2 Protein |
| Purification Method | Protein |
| Immunogen | MPTSSSTKKTQLQLEHLLLDLQMILNGINNYKNPKLTRMLTFKFYMPKKATELKHLQCLEEELKPLEEVLNLAQSKNFHLRPRDLISNINVIVLELKGSETTFMCEYADETATIVEFLNRWITFSQSIISTLT |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
