Learn More
Description
Specifications
Specifications
| Accession Number | P01563.1 |
| Gene ID (Entrez) | 3440 |
| Name | IFN-alpha 2 |
| Purification Method | SDS-PAGE |
| Storage Requirements | Store at -20C. |
| Gene Alias | alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 |
| Gene Symbol | IFNA2 |
| Cross Reactivity | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
