Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Kidney-associated antigen 1 His Protein
SDP

Catalog No. NBP17880110
Click to view available options
:
10μg; Unlabeled

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-84 of Human Kidney-associated antigen 1 The Recombinant Human Kidney-associated antigen 1 Protein is derived from E. coli. The Recombinant Human Kidney-associated antigen 1 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 100mM NaCl
Gene ID (Entrez) 353219
Molecular Weight (g/mol) 11.1kDa
Name Kidney-associated antigen 1 Protein
Purification Method SDS-PAGE
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.