Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human KLF7 His Protein
SDP

Catalog No. NBP223176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223176 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223176 Supplier Novus Biologicals™ Supplier No. NBP223176
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-302 of Human KLF7 The Recombinant Human KLF7 Protein is derived from E. coli. The Recombinant Human KLF7 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003700
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 8609
Molecular Weight (g/mol) 35.8kDa
Name KLF7 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen KLF7, 1-302aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMDVLASY SIFQELQLVH DTGYFSALPS LEETWQQTCL ELERYLQTEP RRISETFGED LDCFLHASPP PCIEESFRRL DPLLLPVEAA ICEKSSAVDI LLSRDKLLSE TCLSLQPASS SLDSYTAVNQ AQLNAVTSLT PPSSPELSRH LVKTSQTLSA VDGTVTLKLV AKKAALSSVK VGGVATAAAA VTAAGAVKSG QSDSDQGGLG AEACPENKKR VHRCQFNGCR KVYTKSSHLK AHQRTHTGEK PYKCSWEGCE WRFARSDELT RHYRKHTGAK PFKCNHCDRC FSRSDHLALH MKRHI
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.